Learn More
Invitrogen™ Mast Cell Chymase Polyclonal Antibody
Código de producto. 16520432
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
16520432 100 μg 100 microgramos
1 options
Código de producto. 16520432 Proveedor Invitrogen™ N.º de proveedor PA597599
Rabbit Polyclonal Antibody
Mast Cell Chymase Polyclonal Antibody for Western Blot, ELISA
This gene product is a chymotryptic serine proteinase that belongs to the peptidase family S1. It is expressed in mast cells and thought to function in the degradation of the extracellular matrix, the regulation of submucosal gland secretion, and the generation of vasoactive peptides. In the heart and blood vessels, this protein, rather than angiotensin converting enzyme, is largely responsible for converting angiotensin I to the vasoactive peptide angiotensin II. Angiotensin II has been implicated in blood pressure control and in the pathogenesis of hypertension, cardiac hypertrophy, and heart failure. Thus, this gene product is a target for cardiovascular disease therapies. This gene maps to 14q11. 2 in a cluster of genes encoding other proteases.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Mast Cell Chymase
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Concentración 3 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
génica CMA1
N.º de referencia del gen P21844, P23946
Alias de gen alpha-chymase; Chymase; chymase 1; chymase 1 preproprotein transcript E; chymase 1 preproprotein transcript I; chymase 1, mast cell; chymase, heart; chymase, mast cell; CMA1; CYH; CYM; mast cell; Mast cell chymase 1; mast cell protease 3; Mast cell protease 5; mast cell protease I; Mast cell protease III; MCP3P; Mcp5; Mcp-5; Mcpt3; Mcpt5; MCT1; mMCP-5; rMCP-3; rMCP-5; rMCP-III
Símbolos de los genes CMA1
Especie del huésped Rabbit
Inmunógeno Recombinant Human Chymase protein (22-247AA).
Método de purificación Protein G
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 1215, 17228
Especies diana Human, Mouse
Contenido y almacenamiento -20°C or -80°C if preferred
Tipo de producto Antibody
Formulario Liquid
Isotype IgG