Learn More
Masitinib, MedChemExpress
Código de producto. 16460097
Encompass_Preferred
Change view
Click to view available options
Cantidad:
10 mg
50 mg
100 mg
200 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
200 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16460097 50 mg 50 miligramos
16493708 10 mg 10 miligramos
16434786 100 mg 100 miligramos
16490946 200 mg 200 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16460097 Proveedor Medchem Express N.º de proveedor HY10209
MedChemExpress Masitinib (AB1010) is a potent, orally bioavailable, and selective inhibitor of c-Kit (IC50=200 nM for human recombinant c-Kit). It also inhibits PDGFRα/β (IC50s=540/800 nM), Lyn (IC50= 510 nM for LynB), Lck, and, to a lesser extent, FGFR3 and FAK. Masitinib (AB1010) has anti-proliferative, pro-apoptotic activity and low toxicity.
Masitinib (AB1010) is a potent, orally bioavailable, and selective inhibitor of c-Kit (IC50=200 nM for human recombinant c-Kit). It also inhibits PDGFRα/β (IC50s=540/800 nM), Lyn (IC50= 510 nM for LynB), Lck, and, to a lesser extent, FGFR3 and FAK. Masitinib (AB1010) has anti-proliferative, pro-apoptotic activity and low toxicity.
Especificaciones
Nombre del producto químico o material Masitinib
Forma física Solid
CAS 790299-79-5
Color Amarillo claro
Cantidad 50 mg
Notas de grado de pureza Research
Fórmula molecular C28H30N6OS
Para utilizar con (aplicación) Cancer-Kinase/protease
Sinónimo AB1010
Información de solubilidad DMSO : 100 mg/mL (200.55 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(NC1=CC=C(C(NC2=NC(C3=CC=CN=C3)=CS2)=C1)C)C4=CC=C(CN5CCN(CC5)C)C=C4
Peso molecular (g/mol) 498.64
Formula Weight (peso de la fórmula) 498.64
Porcentaje de pureza 95.0%