Learn More
STEMCELL Technologies LY364947
Descripción
- LY364947 is a selective inhibitor of the TGFβ/Activin/NODAL pathway that inhibits ALK5 (IC50 = 59 nM)(Sawyer et al.) LY364947 less effectively inhibits TGFβRII (IC50 = 400 nM), p38 MAPK (IC50 = 740 nM), and mixed lineage kinase-7 (MLK-7; IC50 = 1,400 nM) (Li et al., 2006; Sawyer et al.)
Especificaciones
Especificaciones
| CAS | 396129-53-6 |
| Sinónimo | E-616451; HTS 466284; TGF-β RI Kinase Inhibitor |
| Cantidad | 5 mg |
| Fórmula molecular | C17H12N4 |
| Pureza | ≥98% |