Learn More
LY2955303, MedChemExpress
Código de producto. 16490497
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10 miligramos
25 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
16490497 50 mg 50 miligramos
16484148 10 mg 10 miligramos
16466857 5 mg 5 miligramos
16402976 25 mg 25 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16490497 Supplier Medchem Express Supplier No. HY107765
This item is not returnable. View return policy
MedChemExpress LY2955303 is a potent and selective retinoic acid receptor gamma (RARγ) antagonist with a Ki of 1.09 nM.
LY2955303 is a potent and selective retinoic acid receptor gamma (RARγ) antagonist with a Ki of 1.09 nM.
Specifications
Nombre del producto químico o material LY2955303
Forma física Solid
CAS 1433497-19-8
Color Blanco
Cantidad 50 mg
Notas de grado de pureza Research
Fórmula molecular C36H42N4O3
Para utilizar con (aplicación) COVID-19-immunoregulation
Información de solubilidad DMSO : 30 mg/mL (51.84 mM; Need ultrasonic and warming)
SMILES CC(C)(C1=CC(C(C)(C)C)=CC(C2=CC(C3=CC=C(C(O)=O)C=C3)=NN2C4=CC=C(C(N5CCN(C)CC5)=O)C=C4)=C1)C
Peso molecular (g/mol) 578.74
Formula Weight (peso de la fórmula) 578.74
Porcentaje de pureza 99.16%