Learn More
LY2922470, MedChemExpress
Código de producto. 16477736
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16477736 100 mg 100 miligramos
16467938 10 mg 10 miligramos
16463877 50 mg 50 miligramos
16400408 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16477736 Proveedor Medchem Express N.º de proveedor HY19835
MedChemExpress LY2922470 is a potent, selective and orally available agonist of the G protein-coupled receptor 40 (GPR40), with EC50s of 7 nM, 1 nM and 3 nM for human GPR40, mouse GPR40 and rat GPR40, respectively. LY2922470 reduces glucose levels along with significant increases in insulin and GLP-1, is potential for the treatment of type 2 diabetes mellitus (T2DM).
LY2922470 is a potent, selective and orally available agonist of the G protein-coupled receptor 40 (GPR40), with EC50s of 7 nM, 1 nM and 3 nM for human GPR40, mouse GPR40 and rat GPR40, respectively. LY2922470 reduces glucose levels along with significant increases in insulin and GLP-1, is potential for the treatment of type 2 diabetes mellitus (T2DM).
Especificaciones
Nombre del producto químico o material LY2922470
CAS 1423018-12-5
Color Light Yellow
Notas de grado de pureza Research
Fórmula molecular C28H29NO4S
Cantidad 100 mg
Información de solubilidad DMSO : ≥ 125 mg/mL (262.83 mM)
SMILES COC1=C(N(CC2=CC=C(COC3=CC=C([C@@H](C#CC)CC(O)=O)C=C3)S2)CCC4)C4=CC=C1
Peso molecular (g/mol) 475.6
Formula Weight (peso de la fórmula) 475.6
Porcentaje de pureza 98.03%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Metabolism-sugar/lipid metabolism