Learn More
MedChemExpress LY2334737
Product Code. 16418477
Encompass_Preferred
Change view
Click to view available options
Cantidad:
5 mg
Unit Size:
5 miligramos
Product Code. Cantidad unitSize
16418477 5 mg 5 miligramos
1 options
This item is not returnable. View return policy
Product Code. 16418477 Supplier MedChemExpress Supplier No. HY13672
This item is not returnable. View return policy
MedChemExpress LY2334737 is an nucleoside analog and is an orally active prodrug of Gemcitabine. LY2334737 exhibits inhibitory activity against enterovirus A71 (EV-A71) infection. LY2334737 has antiviral and anticancer effects.
Specifications
Peso molecular 389.39 g/mol
Tipo de producto Reference compound
Contenido y almacenamiento Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Condiciones de envío Temperatura ambiente
Pureza 99.02%
Cantidad 5 mg
Información de solubilidad DMSO : ≥ 100 mg/mL (256.81 mM)
Para utilizar con (aplicación) Cancer-programmed cell death
CAS 892128-60-8