Learn More
Tocris Bioscience™ LY 450139
Descripción
LY 450139 is a γ-secretase pseudo-inhibitor. Inhibits Aβ secretion from cells in vitro and increases accumulation of γ-secretase products and long Aβ species in vitro and in vivo. Also attenuates the accumulation of activated microglia and neurological injury in a traumatic brain injury animal model.
Especificaciones
Especificaciones
| Nombre del producto químico o material | (2S)-2-Hydroxy-3-methyl-N-[(1S)-1-methyl-2-oxo-2-[[(1S)-2,3,4,5-tetrahydro-3-methyl-2-oxo-1H-3-benzazepin-1-yl]amino]ethyl]butanamide |
| CAS | 425386-60-3 |
| Cantidad | 5 mg |
| Fórmula molecular | C19H27N3O4 |
| Formula Weight (peso de la fórmula) | 361.44 |
| Pureza | 98% |