Learn More
Tocris Bioscience™ LY 404187
Selective AMPA receptor positive allosteric modulator
Marca: Tocris Bioscience™ 5297/10
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
LY 404187 is a selective AMPA receptor positive allosteric modulator (IC50 values are 0.15, 0.21, 1.66 and 5.65 μM for GluA2i, GluA4i, GluA3i and GluA1i respectively). Potentiates glutamate and AMPA-evoked currents in pre frontal cortex (PFC) pyramidal neurons in vitro. Has no effect on NMDA- or kainate-evoked currents; also has no effect on K+ or Na+ ion channels. Potentiates PFC glutamatergic synaptic transmission in vitro and in vivo. Induces neurite growth in combination with (S)-AMPA (Cat. No. 0254) in SH-SY5Y human neuroblastoma cells in vitro. Also reduces MPTP-induced toxicity in mice. Brain penetrant.Especificaciones
| N-[2-(4′-Cyano[1,1′-biphenyl]-4-yl)propyl]-2-propanesulfonamide | |
| 211311-95-4 | |
| 10 mg |