Learn More
Tocris Bioscience™ LY 2033298
Descripción
LY 2033298 is a selective positive allosteric modulator of M4 receptor (KB = 200 nM, α = 35); increases the potency of acetylcholine by 40-fold. Exhibits no effect at hM1/3/5 receptors and a small effect at hM2 (KB = 1 μM α = 3.7). Potentiates other full and partial orthosteric agonists, including Carbachol, Oxotremorine-M, and McN-A343 . Does not affect binding of antagonist NMS. Active in vivo. Antipsychotic.
α indicates the degree of allosteric enhancement when both orthosteric and allosteric sites are occupied (derived from the allosteric ternary complex model).
Especificaciones
Especificaciones
| Nombre del producto químico o material | 3-Amino-5-chloro-N-cyclopropyl-6-methoxy-4-methylthieno[2,3-b]pyridine-2-carboxamide |
| CAS | 886047-13-8 |
| Cantidad | 5 mg |
| Fórmula molecular | C13H14ClN3O2S |
| Pureza | 98% |