Learn More
Tocris Bioscience™ LW 6
Descripción
LW 6 is a malate dehydrogenase-2 (MDH2) inhibitor (IC50 = 6.3 μM). Also inhibits HIF-1α. Prevents accumulation of HIF-1α in Hep3B cells under hypoxic conditions (IC50 = 2.6 μM) with resulting decrease in expression of HIF-1α target genes VEGF and EPO. Inhibits proliferation of activated human T cells ex vivo.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Methyl 4-hydroxy-3-[[2-(4-tricyclo[3.3.1.13,7 ]dec-1-ylphenoxy)acetyl]amino]benzoate |
| CAS | 934593-90-5 |
| Cantidad | 10 mg |
| Objetivo | Dehydrogenase Inhibitors |
| Fórmula molecular | C26H29NO5 |
| Pureza | 0.98 |