Learn More
LV-320, MedChemExpress
Código de producto. 16487397
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16487397 5 mg 5 miligramos
16434688 10 mg 10 miligramos
16430977 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16487397 Proveedor Medchem Express N.º de proveedor HY112711
MedChemExpress LV-320 is a potent and uncompetitive ATG4B inhibitor with an IC50 of 24.5 μM and a Kd of 16 μM. LV-320 inhibits ATG4B enzymatic activity, blocks autophagic flux in cells, and is stable, non-toxic and active in vivo.
LV-320 is a potent and uncompetitive ATG4B inhibitor with an IC50 of 24.5 μM and a Kd of 16 μM. LV-320 inhibits ATG4B enzymatic activity, blocks autophagic flux in cells, and is stable, non-toxic and active in vivo.
Especificaciones
Nombre del producto químico o material LV-320
Forma física Solid
CAS 2449093-46-1
Color Yellow
Cantidad 5 mg
Notas de grado de pureza Research
Fórmula molecular C29H26ClNO2S2
Para utilizar con (aplicación) Cancer-Kinase/protease
Información de solubilidad DMSO : 135 mg/mL (259.56 mM; Need ultrasonic)
SMILES ClC1=CC2=NC=CC(/C=C/C3=CC=C(C(SCCC(O)=O)SCCC4=CC=CC=C4)C=C3)=C2C=C1
Peso molecular (g/mol) 520.11
Formula Weight (peso de la fórmula) 520.11
Porcentaje de pureza 95.0%