missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LTBR Monoclonal Antibody (C1), Invitrogen™
Descripción
The protein encoded by this gene is a member of the tumor necrosis factor (TNF) family of receptors. It is expressed on the surface of most cell types, including cells of epithelial and myeloid lineages, but not on T and B lymphocytes. The protein specifically binds the lymphotoxin membrane form (a complex of lymphotoxin-alpha and lymphtoxin-beta). The encoded protein and its ligand play a role in the development and organization of lymphoid tissue and tranformed cells. Activation of the encoded protein can trigger apoptosis.
Especificaciones
Especificaciones
| Antígeno | LTBR |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | C1 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| génica | Ltbr |
| N.º de referencia del gen | P36941 |
| Alias de gen | AI256028; CD18; D12S370; LT beta-R; Ltar; LT-beta receptor; LTbetaR; LT-BETA-R; Ltbr; lymphotoxin B receptor; lymphotoxin beta receptor; lymphotoxin beta receptor (TNFR superfamily, member 3); lymphotoxin-beta receptor; TNF receptor-related protein; Tnfbr; TNFCR; TNFR2-RP; TNFR3; TNF-RIII; TNF-R-III; TNFR-III; TNFRrp; TNFR-RP; Tnfrsf3; Tumor necrosis factor C receptor; tumor necrosis factor receptor 2-related protein; Tumor necrosis factor receptor superfamily member 3; tumor necrosis factor receptor superfamily, member 3; tumor necrosis factor receptor type III |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?