missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRRC23 Polyclonal Antibody

Descripción
Recommended positive controls: IMR32. Predicted reactivity: Human (99%), Mouse (84%), Rat (82%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The LRRC23 (Leucine-Rich Repeat Containing 23) gene encodes a protein that is an evolutionarily conserved component of the radial spoke complex in sperm flagella. It is located on human chromosome 12. The LRRC23 protein interacts with radial spoke head proteins such as RSPH3A and RSPH3B, crucial for the structural integrity and proper function of the radial spokes, which are essential for flagellar motility. Mutations or deficiencies in LRRC23 have been linked to male infertility, specifically causing asthenozoospermia, characterized by reduced sperm motility. Studies have identified various mutations in LRRC23 that result in disorganized radial spokes and impaired sperm motility, leading to infertility.
Especificaciones
Especificaciones
| Antígeno | LRRC23 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| génica | LRRC23 |
| N.º de referencia del gen | Q53EV4 |
| Alias de gen | 4921537K05Rik; B7; leucine rich protein, B7; leucine rich repeat containing 23; leucine-rich protein B7; Leucine-rich repeat-containing protein 23; LRPB7; LRRC23; neuron-specific enolase |
| Símbolos de los genes | LRRC23 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?