Learn More
Loxiglumide, Tocris Bioscience™
CCK1 Receptor Antagonists
Brand: Tocris Bioscience 3036/25
This item is not returnable.
View return policy
Description
Loxiglumide is a CCK1 antagonist. Suppresses CCK-induced proliferation of acinar cells. Inhibits pancreatic secretion of digestive enzymes. Active in vivo.Specifications
| 107097-80-3 | |
| 4-[(3,4-Dichlorobenzoyl)amino]-5-[(3-methoxypropyl)pentylamino]-5-oxopentanoic acid | |
| 25 mg |