Learn More
Tocris Bioscience™ Loxiglumide
Descripción
Loxiglumide is a CCK1 antagonist. Suppresses CCK-induced proliferation of acinar cells. Inhibits pancreatic secretion of digestive enzymes. Active in vivo.
Specifications
Specifications
| CAS | 107097-80-3 |
| Sinónimo | 4-[(3,4-Dichlorobenzoyl)amino]-5-[(3-methoxypropyl)pentylamino]-5-oxopentanoic acid |
| Cantidad | 5 mg |
| Fórmula molecular | C21H30Cl2N2O5 |
| Formula Weight (peso de la fórmula) | 461.38 |
| Pureza | 97% |