Learn More
Tocris Bioscience™ Loreclezole hydrochloride
GREENER_CHOICE
Código de producto. 30001229
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
50 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
30001229 10 mg 10 miligramos
30001230 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 30001229 Proveedor Tocris Bioscience™ N.º de proveedor 1295/10
Subtype-selective GABAA modulator
Subtype-selective GABAA receptor modulator. Acts as a positive allosteric modulator of β2 or β3-subunit containing receptors. Also acts as a negative modulator at a novel regulatory site, enhancing GABAA receptor sensitization. Inhibits homomeric ρ1 GABAC receptors.
TRUSTED_SUSTAINABILITY
CAS 2227372-56-5
Fórmula molecular C10H7Cl4N3
Especificaciones
Nombre del producto químico o material Loreclezole hydrochloride
Cantidad 10 mg
Formula Weight (peso de la fórmula) 311
Pureza >99%
Peso molecular 310.987