Learn More
Lonidamine, MedChemExpress
Código de producto. 16498498
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16498498 10 mg 10 miligramos
16400958 5 mg 5 miligramos
16424377 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16498498 Proveedor Medchem Express N.º de proveedor HYB0486
MedChemExpress Lonidamine (AF-1890) is a hexokinase and mitochondrial pyruvate carrier inhibitor (Ki: 2.5 μM). Lonidamine also inhibits aerobic glycolysis in cancer cells. Lonidamine can be used in the research of mitochondrial metabolism and inflammation, such as pulmonary fibrosis.
Lonidamine (AF-1890) is a hexokinase and mitochondrial pyruvate carrier inhibitor (Ki: 2.5 μM). Lonidamine also inhibits aerobic glycolysis in cancer cells. Lonidamine can be used in the research of mitochondrial metabolism and inflammation, such as pulmonary fibrosis.
Especificaciones
Nombre del producto químico o material Lonidamine
Forma física Solid
CAS 50264-69-2
Color Light Nude
Cantidad 10 mg
Notas de grado de pureza Research
Fórmula molecular C15H10Cl2N2O2
Para utilizar con (aplicación) Cancer-Kinase/protease
Sinónimo AF-1890 Diclondazolic Acid DICA
Información de solubilidad DMSO : 50 mg/mL (155.69 mM; Need ultrasonic)
SMILES O=C(C1=NN(CC2=CC=C(Cl)C=C2Cl)C3=C1C=CC=C3)O
Peso molecular (g/mol) 321.16
Formula Weight (peso de la fórmula) 321.16
Porcentaje de pureza 97.08%