Learn More
Tocris Bioscience™ LM11A 31 dihydrochloride
Descripción
LM11A 31 dihydrochloride is a nonpeptide p75NTR ligand and nerve growth factor (NGF) antagonist. LM11A 31 blocks p75-mediated cell death, inhibits proNGF induced cell death (IC50 = 1-10 nM), increases proliferation and survival of hippocampal neural progenitors and inhibits Aβ-induced cell death (EC50 = 20 nM). In animal models of Alzheimer's disease LM11A 31 prevents and reverses atrophy of cholinergic neurites and reverses Alzheimer's pathology; in models of Huntington's disease LM11A 31 increases survival, reduces the formation of aggregates of mutant huntingtin and neuronal degeneration. Also promotes functional recovery in a mouse model of spinal cord injury. Orally available and brain penetrant.
Especificaciones
Especificaciones
| Pureza | 95% |
| Fórmula molecular | (2 S,3 S)-2-Amino-3-methyl-N-[2-(4-morpholinyl)ethyl]pentanamide dihydrochloride |
| CAS | 1243259-19-9 |
| Contenido y almacenamiento | Desiccate at RT |
| Cantidad | 50 mg |
| Tipo de producto | LM11A 31 dihydrochloride |
| Peso molecular | M.W. (Observed): 316.27 |
| Inhibidores | LM11A 31 dihydrochloride |
Para uso exclusivo en investigación