Learn More
Liquiritin, MedChemExpress
Código de producto. 16481388
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
25 miligramos
5 miligramos
50 miligramos
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
16481388 5 mg 5 miligramos
16478958 10 mg 10 miligramos
16459066 100 mg 100 miligramos
16424827 50 mg 50 miligramos
16403866 25 mg 25 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Código de producto. 16481388 Proveedor Medchem Express N.º de proveedor HYN0376
MedChemExpress Liquiritin, a flavonoid isolated from Glycyrrhiza, is a potent and competitive AKR1C1 inhibitor with IC50s of 0.62 μM, 0.61 μM, and 3.72μM for AKR1C1, AKR1C2 and AKR1C3, respectively. Liquiritin efficiently inhibits progesterone metabolism mediated by AKR1C1 in vivo. Liquiritin acts as an antioxidant and has neuroprotective, anti-cancer and anti-inflammatory activity.
Liquiritin, a flavonoid isolated from Glycyrrhiza, is a potent and competitive AKR1C1 inhibitor with IC50s of 0.62 μM, 0.61 μM, and 3.72μM for AKR1C1, AKR1C2 and AKR1C3, respectively. Liquiritin efficiently inhibits progesterone metabolism mediated by AKR1C1 in vivo. Liquiritin acts as an antioxidant and has neuroprotective, anti-cancer and anti-inflammatory activity.
Especificaciones
Nombre del producto químico o material Liquiritin
CAS 551-15-5
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C21H22O9
Cantidad 5 mg
Información de solubilidad DMSO : 150 mg/mL (358.52 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES O=C1C[C@@H](C2=CC=C(O[C@H]3[C@@H]([C@H]([C@@H]([C@@H](CO)O3)O)O)O)C=C2)OC4=CC(O)=CC=C14
Peso molecular (g/mol) 418.39
Formula Weight (peso de la fórmula) 418.39
Porcentaje de pureza 98.3%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-programmed cell death