Learn More
Levomepromazine, MedChemExpress
Código de producto. 16494617
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16494617 50 mg 50 miligramos
16448796 100 mg 100 miligramos
16448718 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16494617 Proveedor Medchem Express N.º de proveedor HYB1693
MedChemExpress Levomepromazine (Methotrimeprazine) is an orally available neuroleptic agent, which is commonly used to relieve nausea and vomiting in palliative care settings. Levomepromazine has antagonist actions at multiple neurotransmitter receptor sites, including dopaminergic, cholinergic, serotonin and histamine receptors.
Levomepromazine (Methotrimeprazine) is an orally available neuroleptic agent, which is commonly used to relieve nausea and vomiting in palliative care settings. Levomepromazine has antagonist actions at multiple neurotransmitter receptor sites, including dopaminergic, cholinergic, serotonin and histamine receptors.
Especificaciones
Nombre del producto químico o material Levomepromazine
CAS 60-99-1
Color Off-White
Notas de grado de pureza Research
Fórmula molecular C19H24N2OS
Cantidad 50 mg
Sinónimo Methotrimeprazine
Información de solubilidad DMSO : 14.29 mg/mL (43.50 mM; Need ultrasonic)
SMILES COC1=CC=C2SC3=C(N(C2=C1)C[C@@H](CN(C)C)C)C=CC=C3
Peso molecular (g/mol) 328.47
Formula Weight (peso de la fórmula) 328.47
Porcentaje de pureza 99.5%
Grado Research
Forma física Powder
Para utilizar con (aplicación) Neuroscience-Neuromodulation