Learn More
Motic LED excitation module TRITC, consist of 525nm LEDlamp and TRITC filter cube
Marca:  Motic 1101011400231
LED excitation module TRITC, consist of 525nm LEDlamp and TRITC filter cube
Código de producto. 17179378
  • ONLINE EXCLUSIVE PRICE
    1525.00€
    1480.00€ / paquete de 1
    Ahorro 45.00€
    2% Descuento
Solo quedan null
Fecha estimada de envoi: 05-06-2026