missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) Nitrate, TRC
Especificaciones
Spécification
| Nombre del producto químico o material | Lead nitrate |
| Cantidad | 10 g |
| Fórmula InChI | InChI=1S/2NO3.Pb/c2*2-1(3)4;/q2*-1;+2 |
| Formula Weight (peso de la fórmula) | 331.9523 |
Nom du produit
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?