Learn More
Tocris Bioscience™ LDN 193188
Inhibitor of phosphatidylcholine transfer protein (PC-TP)
Brand: Tocris Bioscience™ 5631/50
This item is not returnable.
View return policy
Description
LDN 193188 is an inhibitor of phosphatidylcholine transfer protein (PC-TP). Regulates hepatic glucose metabolism. Also promotes insulin-independent phosphorylation of key insulin signaling molecules.Specifications
| 2,4-Dichloro-N-[[[4-[[(4,6-dimethyl-2-pyrimidinyl)amino]sulfonyl]phenyl]amino]carbonyl]benzamide | |
| 1267610-30-9 | |
| 50 mg |