missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LDHA Polyclonal Antibody

Código de producto. 11367693
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
11367693 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11367693 Proveedor Invitrogen™ N.º de proveedor PA519754

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Heat mediated antigen retrieval recommended prior to tissue staining. This antibody is predicted to react with mouse, rat, rabbit, chimpanzee and chinese hamster based on sequence homology.

Lactate dehydrogenase (LDH) is the enzyme in the glycolytic pathway that converts pyruvate to lactate with concomitant interconversion of NADH and NAD+. In mammals, the enzyme is encoded by three genes: LDHA (M or muscle form), LDHB (H or heart form), and LDHC (X or testis form). The three human LDHs have 84-89% sequence similarities and 69-75% amino acid identities. There are five different LDH isoenzymes based on the proportion of M and H chains existing in the LDH tetrameric structure. The LDHA is hypoxia inducible and its expression is directly controlled by the transcriptional activity of the hypoxia inducible factor 1a (HIF1a). LDHB is also known to be upregulated in many cancers. Mutations in the LDHA gene can lead to Glycogen storage disease 11 (GSD11).
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno LDHA
Aplicaciones Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 0.9 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA and 0.02% sodium azide; pH 7.4
génica LDHA
N.º de referencia del gen P00338
Alias de gen Cell proliferation-inducing gene 19 protein; epididymis secretory sperm binding protein Li 133P; GSD11; HEL-S-133P; l7R2; Lactate Dehydrogenase; lactate dehydrogenase 1, A chain; lactate dehydrogenase 3, C chain, sperm specific; lactate dehydrogenase A; lactate dehydrogenase A4; lactate dehydrogenase A-like; lactate dehydrogenase C; lactate dehydrogenase C variant 3; lactate dehydrogenase M; lactate dehydrogenase-A; lactate dehydrogenase-M; lactate dehydrogenase-M (EC 1.1.1.27); Lactic Dehydrogenase; LDH; LDH muscle subunit; LDH testis subunit; Ldh1; Ldh-1; Ldh3; Ldha; LDH-A; ldha protein; ldha1; LDH-A4 muscle-specific isozyme; glycolytic enzyme; Ldhc; LDH-C; Ldhm; LDH-M; LDH-X; L-lactate dehydrogenase; L-lactate dehydrogenase A chain; L-lactate dehydrogenase C chain; M-LDH; OTTHUMP00000165206; PIG19; proliferation-inducing gene 19; renal carcinoma antigen NY-REN-59
Símbolos de los genes LDHA
Especie del huésped Rabbit
Inmunógeno Synthetic peptide conjugated to KLH derived from within residues 300 to the C-terminus of Human Lactate Dehydrogenase.
Método de purificación Antigen Affinity Chromatography
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3939
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.