Learn More
Invitrogen™ LCLAT1 Polyclonal Antibody
GREENER_CHOICE
Código de producto. 13412617
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
13412617 100 μL 100 microlitros
1 options
Código de producto. 13412617 Proveedor Invitrogen™ N.º de proveedor PA528498
Rabbit Polyclonal Antibody
Recommended positive controls: HeLa, HeLa membrane. Predicted reactivity: Mouse (87%), Pig (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Exhibits acyl-CoA:lysocardiolipin acyltransferase (ALCAT) activity; catalyzes the reacylation of lyso-cardiolipin to cardiolipin (CL), a key step in CL remodeling. Recognizes both monolysocardiolipin and dilysocardiolipin as substrates with a preference for linoleoyl-CoA and oleoyl-CoA as acyl donors. Also exhibits 1-acyl-sn-glycerol-3-phosphate acyltransferase activity (AGPAT) activity; converts 1-acyl-sn-glycerol-3- phosphate (lysophosphatidic acid or LPA) into 1,2-diacyl-sn-glycerol-3- phosphate (phosphatidic acid or PA) by incorporating an acyl moiety at the sn-2 position of the glycerol backbone (PubMed:16620771). Possesses both lysophosphatidylinositol acyltransferase (LPIAT) and lysophosphatidylglycerol acyltransferase (LPGAT) activities (PubMed:19075029). Required for establishment of the hematopoietic and endothelial lineages. [UniProt].
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno LCLAT1
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
génica LCLAT1
N.º de referencia del gen Q6UWP7
Alias de gen 1-acylglycerol-3-phosphate O-acyltransferase 8; 1-AGP acyltransferase 8; 1-AGPAT 8; 1AGPAT8; Acyl-CoA:lysocardiolipin acyltransferase 1; Agpat8; AI181996; Alcat1; Gm91; HSRG1849; LCLAT1; Lycat; lysocardiolipin acyltransferase 1; RGD1565906; UNQ1849; UNQ1849/PRO3579
Símbolos de los genes LCLAT1
Especie del huésped Rabbit
Inmunógeno Recombinant protein encompassing a sequence within the center region of human LYCAT. The exact sequence is proprietary.
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 253558
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG