Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-Biotin Antibody Kit
Descripción
The LanthaScreen™ terbium (Tb)-labeled anti-biotin antibody is a generic reagent that can be used to easily label biotinylated biomolecules for use in TR-FRET assays. Although the terbium chelate donor can be directly attached to a biomolecule such as a protein or peptide, it can also be indirectly attached to a target molecule through a biotin-mediated association, in the form of a Tb-labeled streptavidin or anti-biotin antibody. Invitrogen’s Tb-labeled biotin-binding reagents are universal reagents that easily label biotinylated biomolecules with terbium for use in TR-FRET assays for proteases, kinases, etc.
Contents and Storage:
Each LanthaScreen™ Tb-labeled anti-biotin antibody is supplied in HEPES-buffered saline. The kit also includes 400μl of a fluorescein-labeled positive control and 100 ml of TR-FRET dilution buffer. Store components as indicated in the product Certificate of Analysis (-20°C, or +20-30°C).
Contents and Storage:
Each LanthaScreen™ Tb-labeled anti-biotin antibody is supplied in HEPES-buffered saline. The kit also includes 400μl of a fluorescein-labeled positive control and 100 ml of TR-FRET dilution buffer. Store components as indicated in the product Certificate of Analysis (-20°C, or +20-30°C).
Especificaciones
Especificaciones
| Aplicaciones | Kinase Assay, Enzyme and Protein Activity Assays |
| Concentración | 1.1mg/mL |
| Conjugado | Tb (Terbium)/Lanthanide Chelates |
| Cantidad | 25 μg |
| Estado normativo | RUO |
| Disciplina de investigación | TR-FET Assay Development |
| Primario o secundario | Primary |
| Envase | 1 Kit(s) |
| Incluye | 0.1% Sodium azide |
| Contenido y almacenamiento | Store in freezer (-5 to -30°C). |
| Mostrar más |
For Research Use Only. Not for use in diagnostic procedures.