Learn More
GVS Laminated PTFE Membranes
Descripción
- Ideal for vent filtration, phase separation, aerosol sampling
- Prewet with solvent to filter aqueous solutions
- Biologically inert, chemical resistant — chemically compatible with most solvents, acids, alcohols, hydrocarbons, alkalis
- Easy to handle
- PTFE laminated to polypropylene woven support for extra durability
- Thickness: 130μm
- Autoclavable up to 130°C (265°F)
Especificaciones
Especificaciones
| Esterilizables en autoclave | Autoclavable |
| Punto de burbuja | 9psi |
| Color | White |
| Material | PTFE |
| Tamaño de poro | 0.5 μm |
| Para utilizar con (aplicación) | Laminated membranes are ideal for vent filtration, phase separation, aerosol sampling. |
| Caudal de gas | 4.8L/min./cm sq. |
| Hidrofobicidad | Hydrophobic |
| Cantidad | 100/Pk. |
| Grosor (métrico) | 130 μm |
| Mostrar más |