Learn More
LAG-3 (CD223) Mouse anti-Human, PE-CF594, Clone: T47-530, BD Biosciences™
Código de producto. 17267484
Change view
Click to view available options
Cantidad:
25 Tests
Tamaño de la unidad:
25 tests
17267484 25 Tests 25 tests
1 options
Código de producto. 17267484 Proveedor BD Biosciences N.º de proveedor 565719
Mouse Monoclonal Antibody
LAG-3 (CD223) Monoclonal antibody specifically detects LAG-3 (CD223) in Human samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno LAG-3 (CD223)
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon T47-530
Conjugado PE-CF594
Formulación Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
N.º de referencia del gen LAG3; CD223; FDC; Lymphocyte activation gene 3 protein; Protein FDC
Especie del huésped Mouse
Inmunógeno Human LAG-3 Recombinant Protein
Cantidad 25 Tests
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD HorizonTM PE-CF594 under optimum conditions, and unconjugated antibody and free PE-CF594 were removed.
Isotype IgG1 κ