missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD79a/mb-1 (B-Cell Marker) Ab-2 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Código de producto. 12629206 Tienda Epredia Productos
Change view
Click to view available options
Cantidad:
100 μL
200 μL
500 μL
1 mL
7 mL
Formulario:
Concentrated
Prediluted
Concentración:
1mg/mL
200μg/mL
Tamaño de la unidad:
0.50 ml
1 ml
100 microlitros
200 microlitros
7 ml
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad Formulario Concentración unitSize
12629206 500 μL Concentrated 200μg/mL 0.50 ml
12619206 100 μL Concentrated 200μg/mL 100 microlitros
12639206 100 μL Concentrated 1mg/mL 100 microlitros
12649206 200 μL Concentrated 1mg/mL 200 microlitros
12609206 1 mL Concentrated 200μg/mL 1 ml
12679206 7 mL Prediluted - 7 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 12629206 Proveedor Epredia™ N.º de proveedor MS1171P1

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Ensure accurate, reproducible resuts in immunohistochemistry applications with Epredia™ CD79a/mb-1 (B-Cell Marker) Ab-2, Mouse Monoclonal Antibody.

A disulphide-linked heterodimer, consisting of mb-1 (or CD79a) and B29 (or CD79b) polypeptides, is non-covalently associated with membrane-bound immunoglobulins on B cells to constitute the B cell Ag receptor. CD79a first appears at pre B cell stage and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas. It is not present in myeloid or T cell lines.

Host Species: Mouse

Clone: JCB117

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Extracellular domain

Immunogen: Recombinant protein containing part of the extracellular portion of the CD79a glycoprotein

Molecular Weight: 50kDa

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD79a/mb-1 (B-Cell Marker) Ab-2
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon JCB117
Concentración 200μg/mL
Conjugado Unconjugated
Especie del huésped Mouse
Inmunógeno Recombinant protein containing part of the extracellular portion of the CD79a glycoprotein
Cantidad 500 μL
Estado normativo IVD
Disciplina de investigación B-Cell
Primario o secundario Primary
Especies diana Human
Formulario Concentrated
Isotype IgG2a κ
Mostrar más Mostrar menos
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.