Learn More
Tocris Bioscience™ L-817,818
Descripción
Potent and selective somatostatin sst5 receptor agonist. Ki values are 0.4, 3.3, 52, 64 and 82 nM for cloned human sst5, sst1, sst2, sst3 and sst4 receptors respectively. Inhibits growth hormone release from rat pituitary cells (EC50 = 3.1 nM) and insulin release from mouse pancreatic islets (EC50 = 0.3 nM) in vitro.
Especificaciones
Especificaciones
| Nombre del producto químico o material | L-817, 818 |
| CAS | 217480-27-8 |
| Cantidad | 10 mg |
| Fórmula molecular | C33H36N4O3 |
| Formula Weight (peso de la fórmula) | 536.67 |
| Pureza | >99% |
| Peso molecular | 536.676 |