Learn More
Tocris Bioscience™ L-161,982
Descripción
L-161,982 is an EP4 receptor antagonist that is selective over all other members of the prostanoid receptor family (Ki values are 0.024, 0.71, 1.90, 5.10, 5.63, 6.74, 19 and 23 μM for human EP4, TP, EP3, DP, FP, IP, EP1 and EP2 receptors respectively). Suppresses PGE2-induced bone formation in rats and prevents the nociceptive response induced by Misoprostol (Cat. No. 2297) in formalin-injected mice.
Especificaciones
Especificaciones
| Nombre del producto químico o material | L-161, 982 |
| CAS | 147776-06-5 |
| Cantidad | 10 mg |
| Fórmula molecular | C32H29F3N4O4S2 |
| Formula Weight (peso de la fórmula) | 654.72 |
| Pureza | >99% |
| Peso molecular | 654.723 |