Learn More
L 152804, MedChemExpress
Código de producto. 16470497
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16470497 50 mg 50 miligramos
16464148 10 mg 10 miligramos
16446857 5 mg 5 miligramos
16425116 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16470497 Proveedor Medchem Express N.º de proveedor HY107734
MedChemExpress L 152804 is an orally active and selective neuropeptide Y Y5 receptor (NPY5-R) antagonist, with a Ki of 26 nM for hY5. L 152804 causes weight loss in diet-induced obese mice by modulating food intake and energy expenditure.
L 152804 is an orally active and selective neuropeptide Y Y5 receptor (NPY5-R) antagonist, with a Ki of 26 nM for hY5. L 152804 causes weight loss in diet-induced obese mice by modulating food intake and energy expenditure.
Especificaciones
Nombre del producto químico o material L 152804
CAS 6508-43-6
Color Light Yellow
Notas de grado de pureza Research
Fórmula molecular C23H26O4
Cantidad 50 mg
Información de solubilidad DMSO : ≥ 125 mg/mL (341.11 mM)
SMILES O=C1C2=C(CC(C)(C)C1)OC3=CC=CC=C3C2C4C(CC(C)(C)CC4=O)=O
Peso molecular (g/mol) 366.45
Formula Weight (peso de la fórmula) 366.45
Porcentaje de pureza 98.17%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Metabolism-protein/nucleotide metabolism