missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KV3.2 (KCNC2) Mouse anti-Human, Mouse, Rat, Clone: S410-17, Invitrogen™
Descripción
Voltage-gated K+ channels in the plasma membrane control the repolarization and the frequency of action potentials in neurons, muscles and other excitable cells. The KV gene family encodes more than 30 proteins that comprise the subunits of the K+ channels. The subunits vary in their gating and permeation properties, subcellular distribution and expression patterns. Functional KV channels assemble as tetramers consisting of pore-forming alpha subunits, which include the KV1, KV2, KV3, KV4 and KV9 proteins, as well as accessory subunits that modify the gating properties of the coexpressed KV subunits. KV3.2 is a multipass membrane protein that regulates the voltage-dependent K+ permeability of excitable membranes. The tail of KV3.2 may be influential in the targeting of the channel to specific subcellular compartments and/or the regulation of channel activity.
Especificaciones
Especificaciones
| Antígeno | KV3.2 (KCNC2) |
| Aplicaciones | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot |
| Clasificación | Monoclonal |
| Clon | S410-17 |
| Concentración | 1 mg/ml |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.4 |
| génica | KCNC2 |
| N.º de referencia del gen | P22462, Q14B80, Q96PR1 |
| Alias de gen | Potassium voltage-gated channel subfamily C member 2, Kcnc2, KSHIIIA |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?