Learn More
KU-57788, MedChemExpress
Código de producto. 16474348
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16474348 10 mg 10 miligramos
16440677 50 mg 50 miligramos
16427067 5 mg 5 miligramos
16415266 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16474348 Proveedor Medchem Express N.º de proveedor HY1100610mg
MedChemExpress KU-57788 (NU7441) is a highly potent and selective DNA-PK inhibitor with an IC50 of 14 nM. KU-57788 is an NHEJ pathway inhibitor. KU-57788 also inhibits PI3K and mTOR with IC50s of 5.0 and 1.7 μM, respectively.
KU-57788 (NU7441) is a highly potent and selective DNA-PK inhibitor with an IC50 of 14 nM. KU-57788 is an NHEJ pathway inhibitor. KU-57788 also inhibits PI3K and mTOR with IC50s of 5.0 and 1.7 μM, respectively.
Especificaciones
Nombre del producto químico o material KU-57788
CAS 503468-95-9
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C25H19NO3S
Cantidad 10 mg
Sinónimo NU7441
Información de solubilidad DMSO : 14.29 mg/mL (34.56 mM; Need ultrasonic)
SMILES O=C1C=C(N2CCOCC2)OC3=C1C=CC=C3C4=CC=CC5=C4SC6=C5C=CC=C6
Peso molecular (g/mol) 413.49
Formula Weight (peso de la fórmula) 413.49
Porcentaje de pureza 98.85%
Grado Research
Forma física Powder
Para utilizar con (aplicación) Cancer-Kinase/protease