Descripción
- The hydrophobic spunbond of the shoe cover is made from 90% recycled-origin material
- The PE sole incorporates at least 50% recycled-origin material
- The PE sole ensures good mechanical resistance while providing excellent floor adhesion
- Their anatomical shapes allow them to adapt to all types of shoes and morphologies, especially when used without shoes, notably in the medical field
Especificaciones
Especificaciones
| Tamaño | X-Large |
| Material | Polyethylene, Polypropylene |
| Latex Free | Yes |
| Color | White, Blue |
| Desechables | Polyethylene |
| Esterilidad | Non-sterile |
| Tipo de tapón | Elastic Ankle |
| Tipo de suela | Anti-slip |
| Resistencia | Liquid |
| Tipo de producto | Shoe Covers |