missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KNTC1 Polyclonal Antibody

Código de producto. 15584372
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15584372 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. View return policy
Product Code. 15584372 Supplier Invitrogen™ Supplier No. PA540762

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: MWNDIELLTN DDTGSGYLSV GSRKEHGTAL YQVDLLVKIS SEKASLNPKI Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 100%.

This gene encodes a protein that is one of many involved in mechanisms to ensure proper chromosome segregation during cell division.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno KNTC1
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.5 mg/mL
Conjugado Unconjugated
Formulación PBS with 2% sucrose and 0.09% sodium azide
génica KNTC1
N.º de referencia del gen P50748
Alias de gen D330012D13; FLJ36151; HsROD; Kiaa0166; kinetochore associated 1; kinetochore-associated protein 1; Kinetochore-associated protein 1-like protein; KNTC1; LOW QUALITY PROTEIN: kinetochore-associated protein 1; ROD; Rough deal homolog; Rough Deal homolog, centromere/kinetochore protein
Símbolos de los genes KNTC1
Especie del huésped Rabbit
Inmunógeno Synthetic peptide directed towards the N-terminal of human KNTC1 (aa 1-50).
Método de purificación Affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 9735
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.