Learn More
Tocris Bioscience™ Klotho-derived peptide 6
Descripción
Klotho-derived peptide 6 is a β-catenin signaling inhibitor. Binds to and sequesters Wnt ligands in vitro; prevents expression of Wnt1-induced fibronectin and α-smooth muscle action. In an in vivomouse model of diabetic kidney disease, Klotho-derived peptide 6 reduces kidney injury, improves survival and prevents accumulation of fibronectin and collagen IV in glomeruli.
Especificaciones
Especificaciones
| Gene ID (Entrez) | 1499 |
| Pureza | 0.95 |
| CAS | 2102414-23-1 |
| Contenido y almacenamiento | Store at -20°C |
| Alias de gen | bCatenin, beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, CTNNB1, DKFZp686D02253, FLJ25606, FLJ37923, Int-1, KL, Wnt1 |
| Peso molecular | MolecularWeight-observed: 3491.87 |
| Cantidad | 1 mg |