missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KIR2DS2 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 13465987
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
13465987 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 13465987 Proveedor Invitrogen™ N.º de proveedor PA531464

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, Jurkat, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Killer cell immunoglobulin-like receptors (KIRs) are transmembrane glycoproteins expressed by natural killer cells and subsets of T cells. The KIR genes are polymorphic and highly homologous and they are found in a cluster on chromosome 19q13.4 within the 1 Mb leukocyte receptor complex (LRC). The gene content of the KIR gene cluster varies among haplotypes, although several framework genes are found in all haplotypes (KIR3DL3, KIR3DP1, KIR3DL4, KIR3DL2). The KIR proteins are classified by the number of extracellular immunoglobulin domains (2D or 3D) and by whether they have a long (L) or short (S) cytoplasmic domain. KIR proteins with the long cytoplasmic domain transduce inhibitory signals upon ligand binding via an immune tyrosine-based inhibitory motif (ITIM), while KIR proteins with the short cytoplasmic domain lack the ITIM motif and instead associate with the TYRO protein tyrosine kinase binding protein to transduce activating signals. The ligands for several KIR proteins are subsets of HLA class I molecules; thus, KIR proteins are thought to play an important role in regulation of the immune response.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno KIR2DS2
Aplicaciones Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 0.96 mg/mL
Conjugado Unconjugated
Formulación 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
génica KIR2DS2
N.º de referencia del gen P43631
Alias de gen 1060P11.9.2; 183ActI; CD158 antigen-like family member J; CD158b; CD158J; cl-49; killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 2; killer cell immunoglobulin-like receptor 2DS2; killer cell immunoglobulin-like receptor KIR2DS2; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 2; killer-cell Ig-like receptor; killer-cell immunoglobulin-like receptor two domains short tail 2 protein; KIR2DS2; KIR-2DS2; MHC class I NK cell receptor; na; natural killer associated transcript 5; natural killer cell inhibitory receptor; natural killer cell receptor; Natural killer-associated transcript 5; NK receptor 183 ActI; NKAT5; NKAT-5; p58 killer cell inhibitory receptor KIR-K7a; p58 natural killer cell receptor clone CL-49; p58 NK receptor CL-49
Símbolos de los genes KIR2DS2
Especie del huésped Rabbit
Inmunógeno Recombinant protein encompassing a sequence within the center region of human KIR2DS2. The exact sequence is proprietary.
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 100132285
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.