Learn More
Tocris Bioscience™ Ki 8751
Descripción
Ki 8751 is a potent, selective inhibitor of VEGFR-2 tyrosine kinase (IC50 = 0.9 nM). Displays some inhibitory activity towards c-Kit, PDGFRα and FGFR-2 (IC50 values range from 40 to 170 nM) but is highly selective over other receptor tyrosine kinases (IC50 > 10000 nM for FGFR-2, EGFR and HGFR). Inhibits VEGF-stimulated proliferation of human umbilical vein endothelial cells (HUVEC) and inhibits tumor growth in vivo; antiangiogenic.
Especificaciones
Especificaciones
| Nombre del producto químico o material | N-(2,4-Difluorophenyl)-N'-[4-[(6,7-dimethoxy-4-quinolinyl)oxy]-2-fluorophenyl]urea |
| CAS | 228559-41-9 |
| Intervalo de porcentaje de ensayo | >97% |
| Cantidad | 50 mg |
| Solubilidad | Soluble to 100mM in DMSO |
| Fórmula molecular | C24H18F3N3O4 |
| Formula Weight (peso de la fórmula) | Observed MW: 469.41 |
Para uso exclusivo en investigación