Learn More
Tocris Bioscience™ KI-7
Descripción
KI-7 is an adenosine A2B receptor positive allosteric modulator. Potentiates the NECA (Cat. No. 1691) -induced increase in cAMP production in CHO cells expressing A2B receptors. Also potentiates the effects of NECA and the A2B receptor agonist BAY 60-6583 (Cat. No. 4472) in mediating osteoblast differentiation from MSCs in vitro.
Especificaciones
Especificaciones
| Nombre del producto químico o material | α-Oxo-N-phenyl-1-(phenylmethyl)-1H-indole-3-acetamide |
| CAS | 1489263-00-4 |
| Cantidad | 10 mg |
| Fórmula molecular | C23H18N2O2 |
| Formula Weight (peso de la fórmula) | 354.4 |
| Pureza | 98% |