Learn More
Ki 16425
Descripción
Ki 16425 is an antagonist of the lysophosphatidic acid receptors LPA1 and LPA3 (Ki values are 0.25 and 0.36 μM respectively, in a GTPγS binding assay). Blocks LPA-induced dephosphorylation of Yes-associated protein (YAP) and WW domain-containing transcription regulator protein 1 (TAZ), inhibiting the Hippo signaling pathway, in HEK293A cells.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 3-[[[4-[4-[[[1-(2-Chlorophenyl)ethoxy]carbonyl]amino]-3-methyl-5-isoxazolyl]phenyl]methyl]thio]propanoic acid |
| CAS | 355025-24-0 |
| Intervalo de porcentaje de ensayo | >98% |
| Cantidad | 25 mg |
| Fórmula molecular | C23H23ClN2O5S |
| Formula Weight (peso de la fórmula) | Observed MW: 474.96 |