missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KCTD11 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 17886822
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17886822 100 μg 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17886822 Proveedor Invitrogen™ N.º de proveedor PA5142626

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.

Calponin, a thin filament associated protein is implicated in the regulation and modulation of smooth muscle contraction. It is capable of binding to actin, calmodulin, troponin C and tropomyosin. Calponin is expressed in smooth muscle and tissues containing significant amounts of smooth muscle. Two isoforms of calponin exist whose molecular weights are 34kDa and 29kDa. Expression of the 29kDa form is primarily restricted to muscle of the urogenital tract. The expression of calponin has also been demonstrated in myoepithelial cells from benign and malignant breast lesions. It stains smooth muscle, myoepithelial cells, myofibroblasts, keratinocytes and nerve fibers. It identifies myoepithelial cells in breast lesions, and helps differentiate breast collagenous spherulosis (positive) from adenoid cystic carcinoma. Adenoid cystic carcinoma in salivary gland tumors is calponin positive.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno KCTD11
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.5 mg/mL
Conjugado Unconjugated
Formulación TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
génica KCTD11
N.º de referencia del gen Q693B1
Alias de gen AF465352; BTB/POZ domain-containing protein KCTD11; C17orf36; KCASH1; KCASH1 protein; KCTD11; potassium channel tetramerisation domain containing 11; potassium channel tetramerization domain containing 11; potassium channel tetramerization domain-containing protein 11; REN; REN/KCTD11; retinoic acid, EGF, and NGF upregulated; retinoic acid, EGF, NGF induced gene protein; retinoic acid, EGF, NGF induced gene/potassium channel tetramerization domain containing 11; RING-type E3 ubiquitin transferase KCTD11; RING-type E3 ubiquitin transferase subunit KCTD11
Símbolos de los genes KCTD11
Especie del huésped Goat
Inmunógeno Peptide with sequence C-PEVEYGRLGLQP.
Método de purificación Ammonium sulfate precipitation
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 147040
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.