missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KCNMB1 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 11598601
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
11598601 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11598601 Proveedor Invitrogen™ N.º de proveedor PA1924

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

PA1-924 detects large conductance, calcium-activated potassium channel beta (Maxi K+ Beta) from human, mouse, and rat samples. PA1-924 has been successfully used in western blot and immunofluorescence. By western blot, this antibody detects an ~28kDa protein representing Maxi K+ beta from mouse myometrial smooth muscle cell extracts. The molecular weight detected varies due to glycosylation, multimerization and binding to actin. Immunizing peptide corresponds to amino acid residues 90-103 from human Maxi K+ beta. This sequence is completely conserved across many species. PA1-924 immunizing peptide (Cat. # PEP-119) is available for use in neutralization and control experiments.

Potassium channels are a group of ubiquitously expressed proteins that serve numerous functions in excitable and non-excitable cells. One class of integral membrane potassium channels is the large conductance, calcium-activated potassium channel (Maxi K+). Maxi K+ differs from most other potassium channels in that its activation is controlled by both increases in intracellular calcium and by membrane depolarization. Maxi K+ dual activation is possible because of its structure. The core of the channel, which is similar to other potassium channels, is a Maxi K+ alpha homotetramer that contains both a voltage sensor and an intracellular calcium binding domain. In vascular smooth muscle, an auxiliary beta-subunit is found in a 1:1 stoichiometry. The beta-subunit exhibits its effect on the Maxi K+ channel by effectively decreasing by 5- to 10- fold the concentration of calcium required to keep the pore open. Maxi K+ beta is the target for possible therapeutics because of its role in blood flow and blood pressure regulation.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno KCNMB1
Aplicaciones Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1mg/mL BSA and 0.05% sodium azide
génica KCNMB1
N.º de referencia del gen P97678, Q16558, Q8CAE3
Alias de gen big potassium channel beta subunit 1; BK channel beta subunit; BK channel beta subunit 1; BK channel subunit beta-1; BKbeta; BKbeta1; calcium-activated potassium channel beta subunit; calcium-activated potassium channel subunit beta; Calcium-activated potassium channel subunit beta-1; calcium-activated potassium channel, subfamily M subunit beta-1; Charybdotoxin receptor subunit beta-1; hbeta1; hslo-beta; K(VCA)beta; k(VCA)beta-1; Kcnmb1; large conductance Ca2+-activated K+ channel beta 1 subunit; maxi K channel subunit beta-1; MaxiK channel beta-subunit 1; potassium calcium-activated channel subfamily M regulatory beta subunit 1; potassium channel subfamily M regulatory beta subunit 1; potassium large conductance calcium-activated channel, subfamily M, beta member 1; slo-beta; slo-beta-1; slowpoke-beta
Símbolos de los genes KCNMB1
Especie del huésped Rabbit
Inmunógeno Synthetic Peptide: L(90) Y H T E D T R D Q N Q Q C(103).
Método de purificación Antigen affinity chromatography
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 16533, 29747, 3779
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.