Learn More
Tocris Bioscience™ K 252a
Descripción
Non-selective protein kinase inhibitor; analog of staurosporine (Cat.No. 1285). Inhibits PKC (IC50 = 32.9 nM), Ca2+/calmodulin-stimulated phosphodiesterases (IC50 = 1.3 - 2.9 μM), MLCK (Ki = 20 nM) and receptor tyrosine kinases. Inhibits the oncogenic properties of MET; prevents autophosphorylation and activation of downstream effectors (MAPK, Akt).
Especificaciones
Especificaciones
| Nombre del producto químico o material | K 252a |
| CAS | 99533-80-9 |
| Cantidad | 200 μg |
| Fórmula molecular | C27H21N3O5 |
| Formula Weight (peso de la fórmula) | 467.48 |
| Pureza | >98% |
| Peso molecular | 467.481 |