Learn More
Tocris Bioscience™ K 22
Descripción
K 22 is a viral RNA synthesis inhibitor; targets membrane-bound coronaviral RNA replication by inhibiting double membrane vesicle (DMV) formation. Inhibits HCoV-229E infection of MRC-5 cells in a plaque forming assay (IC50 = 0.7 μM). Also inhibits HCoV-229E and MERS-CoV replication in human airway epithelia cultures.
Especificaciones
Especificaciones
| Nombre del producto químico o material | N-[(1 Z)-1-[[4-(4-Bromophenyl)-4-hydroxy-1-piperidinyl]carbonyl]-2-phenylethenyl]benzamide |
| CAS | 2141978-86-9 |
| Cantidad | 50 mg |
| Objetivo | DNA, RNA and Protein Synthesis Inhibitors |
| Fórmula molecular | C27H25BrN2O3 |
| Pureza | 0.98 |