Learn More
Tocris Bioscience™ JX 06
Descripción
JX 06 is a potent and selective pyruvate dehydrogenase kinase (PDK) 1/2/3 inhibitor (IC50 values are 49, 101 and 313 nM for PDK1, PDK2 and PDK3, respectively). Exhibits selectivity for PDK1/2/3 over PDK4 and a panel of 323 other kinases. Suppresses A549 lung cancer cell proliferation in vitro. Reduces tumor volume of A549 xenografts in mice.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Bis(morpholinothiocarbonyl) disulfide |
| CAS | 729-46-4 |
| Cantidad | 50 mg |
| Objetivo | PDK-1 Inhibitors |
| Fórmula molecular | C10H16N2O2S4 |
| Formula Weight (peso de la fórmula) | 324.51 |
| Pureza | >98% |