Learn More
Tocris Bioscience™ JW 74
Wnt pathway inhibitor
Marca: Tocris Bioscience™ 5653/50
This item is not returnable.
View return policy
Description
JW 74 is an inhibitor of canonical Wnt signaling (IC50 = 790 nM). Targets the β-catenin destruction complex (GSK-3?/AXIN/APC). Increases AXIN2 and decreases active β-catenin levels in colorectal cancer (CRC) cells. Also arrests cell cycle in G1 and inhibits proliferation of CRC cells in vitro. Inhibits tumor growth and formation in mouse models.Specifications
| 4-[4-(4-Methoxyphenyl)-5-[[[3-(4-methylphenyl)-1,2,4-oxadiazol-5-yl]methyl]thio]-4H-1,2,4-triazol-3-yl]-pyridine | |
| 863405-60-1 | |
| 50 mg | |
| Soluble to 100mM in DMSO |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only