Learn More
JW 55, Tocris Bioscience™
Código de producto. 18074732
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
50 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18074732 10 mg 10 miligramos
18045522 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18074732 Proveedor Tocris Bioscience N.º de proveedor 4514/10
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos
Tankyrase inhibitor
Inhibitor of the PARP domain of tankyrase 1 and 2 (TNKS1/2). Activity results in stabilization of Axin2 and increased degradation of β-catenin; inhibits the β-catenin signaling pathway. Also shown to decrease canonical Wnt signaling in SW480 and HCT-15 colon carcinoma cell lines; reduces cell cycle progression and proliferation in SW480 cells in vitro.
TRUSTED_SUSTAINABILITY
CAS 664993-53-7
Fórmula molecular C25H26N2O5
Peso molecular (g/mol) 434.49
Número MDL MFCD03863265
Clave InChI ZJZWZIXSGNFWQQ-UHFFFAOYSA-N
Sinónimo n-4-4-4-methoxyphenyl oxan-4-yl methylcarbamoyl phenyl furan-2-carboxamide, n-4-4-4-methoxyphenyl tetrahydro-2h-pyran-4-yl methyl carbamoyl phenyl furan-2-carboxamide, qcr-64, n-4-4-4-methoxyphenyl oxan-4-yl methyl carbamoyl phenyl furan-2-carboxamide, n-4-4-4-methoxyphenyl tetrahydro-2h-pyran-4-ylmethylcarbamoyl phenyl furan-2-carboxamide, n-4-4-4-methoxyphenyl tetrahydro-2h-pyran-4-yl methylcarbamoyl phenyl furan-2-carboxamide, n-4-4-4-methoxyphenyl tetrahydro-2h-pyran-4-yl methyl amino carbonyl phenyl-2-furamide, n-4-tetrahydro-4-4-methoxyphenyl-2h-pyran-4-yl methyl amino carbonyl phenyl-2-furancarboxamide
PubChem CID 2946601
Nombre IUPAC N-[4-({[4-(4-methoxyphenyl)oxan-4-yl]methyl}carbamoyl)phenyl]furan-2-carboxamide
SMILES COC1=CC=C(C=C1)C1(CNC(=O)C2=CC=C(NC(=O)C3=CC=CO3)C=C2)CCOCC1
Especificaciones
Nombre del producto químico o material JW 55
Cantidad 10 mg
Formula Weight (peso de la fórmula) 434.48
Porcentaje de pureza >98%