Learn More
JSH-23, MedChemExpress
Código de producto. 16455948
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16455948 10 mg 10 miligramos
16448577 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16455948 Proveedor Medchem Express N.º de proveedor HY13982
MedChemExpress JSH-23 is an NF-κB inhibitor which inhibits NF-κB transcriptional activity with an IC50 of 7.1 μM in lipopolysaccharide (LPS)-stimulated macrophages RAW 264.7. JSH-23 inhibits nuclear translocation of NF-κB p65 without affecting IκBα degradation.
JSH-23 is an NF-κB inhibitor which inhibits NF-κB transcriptional activity with an IC50 of 7.1 μM in lipopolysaccharide (LPS)-stimulated macrophages RAW 264.7. JSH-23 inhibits nuclear translocation of NF-κB p65 without affecting IκBα degradation.
Especificaciones
Nombre del producto químico o material JSH-23
Forma física Solid
CAS 749886-87-1
Color Purple
Cantidad 10 mg
Notas de grado de pureza Research
Fórmula molecular C16H20N2
Para utilizar con (aplicación) Cancer-programmed cell death
Información de solubilidad DMSO : ≥ 56 mg/mL (233.00 mM)
SMILES NC1=CC(C)=CC=C1NCCCC2=CC=CC=C2
Peso molecular (g/mol) 240.34
Formula Weight (peso de la fórmula) 240.34
Porcentaje de pureza 99.11%