Learn More
Tocris Bioscience™ JQEZ5
Descripción
JQEZ5 is a potent and selective SAM-competitive EZH2 lysine methyltransferase inhibitor (IC50 = 11 nM). Selectively binds EZH2 over a panel of 22 methyltransferases. Reduces global level of the PRC2-associated mark H3K27me3 in K562 cells. Suppresses cell growth in CML cells. Inhibits colony formation of primary human CD34+ CML stem/ progenitor cells. Antitumor effects in vivo.
Especificaciones
Especificaciones
| Nombre del producto químico o material | N-[(1,2-Dihydro-6-methyl-2-oxo-4-propyl-3-pyridinyl)methyl]-1-(1-methylethyl)-6-[6-(4-methyl-1-piperazinyl)-3-pyridinyl]-1H-pyrazolo[3,4-b]pyridine-4-carboxamide |
| CAS | 1913252-04-6 |
| Cantidad | 10 mg |
| Fórmula molecular | C30H38N8O2 |
| Pureza | 98% |